About this item
Whatman GD/X Syringe Filters are high-viscosity filters that incorporate a prefilter to enable filtration of viscous, dense or otherwise hard-to-filter material with high solids content as in environmental and food analysis samples.
- Recyclable after use
Non sterile Whatman GD/X Syringe Filters combine a polypropylene housing, high-efficiency Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:
- Polyethersulfone (PES) for serum, plasma and tissue culture
- Polyvinylidene difluoride (PVDF) for HPLC
- Cellulose acetate for biological solutions
- Borosilicate glass microfiber
The prefilter contains a high-efficiency borosilicate glass filtration stack that will retain fine particles down to 0.7 μm, allowing difficult sample filtration with less hand force. It reduces the need for multiple syringe filters by processing three to seven times more sample volume than syringe filters without prefilters. The filters can be autoclaved.
Cytiva also offers Sterile Whatman GD/X syringe filters.
Specifications
- Catalog No:
- 514-0534
- 514-0535
- 514-0532
- 514-8045
- 514-8046
- 514-0533
- 514-0536
- 514-8064
- 516-1372
- 516-6312
- 516-6313
- 514-8041
- 514-8042
- 514-8043
- 514-0531
- 514-8044
- 516-6314
- 516-6315
- 514-0291
- 514-0292
- 514-8062
- 514-8040
- 514-8063
- 514-8060
- 514-8039
- 514-0528
- 514-0529
- 516-6324
- 516-1373
- 516-0414
- 514-0290
- 516-0990
- 516-0992
- WHAT6871-2550
- 514-0530
- 514-8051
- 514-8052
- 516-6322
- 514-8055
- 514-8054
- 511-0161
- 511-0162
- 516-6317
- 514-8047
- 514-8048
- 516-6318
- 516-6319
- 516-6309
- 514-0680
- 516-6320
- 516-6321
- 514-8049
- 514-8050
- Ø:
- 13 mm
- 25 mm
- 13 mm
- 25 mm
- 25 mm
- 25 mm
- 13 mm
- 25 mm
- 25 mm
- 13 mm
- 13 mm
- 25 mm
- 25 mm
- 13 mm
- 25 mm
- 13 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 13 mm
- 25 mm
- 13 mm
- 13 mm
- 25 mm
- 13 mm
- 25 mm
- 13 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 13 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 13 mm
- 13 mm
- 13 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 13 mm
- 13 mm
- 25 mm
- 25 mm
- Dimensions:
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- 21,6×29,8 mm
- Max. operating pressure:
- 6,9 bar
- 5,2 bar
- 6,9 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 6,9 bar
- 5,2 bar
- 5,2 bar
- 6,9 bar
- 6,9 bar
- 5,2 bar
- 5,2 bar
- 6,9 bar
- 5,2 bar
- 6,9 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 6,9 bar
- 5,2 bar
- 6,9 bar
- 6,9 bar
- 5,2 bar
- 6,9 bar
- 5,2 bar
- 6,9 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 6,9 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 6,9 bar
- 6,9 bar
- 6,9 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 6,9 bar
- 6,9 bar
- 5,2 bar
- 5,2 bar
- Weight:
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
- 3 g
Specifications
Related Information
Certificates
Search by Lot Number28420489_INSTRUCTIONS.pdf 28420490_INSTRUCTIONS 28420491_INSTRUCTIONS.pdf 28420492_INSTRUCTIONS.pdf 28420493_INSTRUCTIONS.pdf