Order Entry
Austria
Orders LinkContactUsLinkComponent
Filter holder, SolVac®

Filter holder, SolVac®

47 mm
Catalog # 514-4001
Filter holder, SolVac®
Catalog # 514-4001
Filter holder, SolVac®
Filter holder, SolVac®

47 mm

Price
542,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Simplifies clean-up and degassing of mobile phase solvents and other solutions. The versatile design fits most HPLC bottles, flasks, and containers, and eliminates the added steps of washing flasks and transferring mobile phase solvent from flask to reservoir.

  • Draws directly from HPLC solvent bottle. Less likely to spill aggressive solvents than glass funnels or disposable cups
  • Patented magnetic seal is reliable and leak proof. Eliminates the possibility of membrane shifting or tearing which can occur with aluminium clamps or threaded holders

Reusable, chemically resistant PP construction is resistant to common HPLC mobile phase solvents such as methanol, acetonitrile, and tetrahydrofuran.

Applications: Remove contaminating particulate from mobile phase or other solutions, de-gas mobile phase solvents and solutions, eliminate pour-and-wait filtration, remove microbial growth every 24 hours from aqueous buffer mobile phases.

Delivery information: Supplied with 610 mm feedline tubing, thumb clamp, sinker, vacuum port adapter, two membrane seal gaskets and two seal gaskets.

Documents

Specifications
  • For Ø:
    47 mm
  • Filtration area:
    10,2 cm²
  • Cat. No.:
    514-4001
  • Inlet/outlet connections:
    Tapered inlet accepts 3,2 - 6,4 mm ID tubing; outlet seals to bottles with openings 17,8 mm ID to 48,3 mm OD
  • Max. operating temperature:
    38 °C
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Sinker replacement kit
    Sinker replacement kit
    516-7571
    Log in for availability
    1 * 2 ST - 70,10 €

Product Information

  • Catalog #
    Availability
    Price
  • Clamp replacement kit
    Clamp replacement kit
    516-7572
    Log in for availability
    1 * 2 ST - 44,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Membrane seal gasket
    Membrane seal gasket
    514-4100
    Log in for availability
    1 * 10 ST - 48,20 €

Product Information

  • Catalog #
    Availability
    Price
  • 1220 mm replacement feedline tubing
    1220 mm replacement feedline tubing
    516-7568
    Log in for availability
    1 * 1 ST - 85,50 €

Product Information

  • Catalog #
    Availability
    Price
  • Flask seal gasket
    Flask seal gasket
    514-4099
    Log in for availability
    1 * 10 ST - 49,90 €

Product Information

  • Catalog #
    Availability
    Price
  • Vacuum port adaptor
    Vacuum port adaptor
    516-1226
    Log in for availability
    1 * 5 ST - 31,40 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.