Filter discs
: KARTELL
HDPE filter discs for use between filter paper and perforated Büchner plate to prevent clogging of filter holes.
HDPE filter discs for use between filter paper and perforated Büchner plate to prevent clogging of filter holes.
Product Family Options
- Ø