Order Entry
Austria
ContactUsLinkComponent
Bottle top vacuum filtration systems, PES membrane, Rapid-Flow™ Nalgene®
Bottle top vacuum filtration systems, PES membrane, Rapid-Flow™ Nalgene®
Catalog # 514-0590
   Low Manufacturing Impact
undefined
Bottle top vacuum filtration systems, PES membrane, Rapid-Flow™ Nalgene®
Catalog # 514-0590
   Low Manufacturing Impact

Specifications

  • Pore size:
    0,20 µm
  • Capacity:
    50 ml
  • Seq:
    02
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Membrane Ø:
    50 mm
  • Pk:
    12

Specifications

About this item

Rapid-Flow™ disposable filter units with PES membrane, PS funnel and PP collection tube feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination and optimal choice for small volume tissue culture work. Offers clean, fast filtration of 20 to 50 ml of media or buffers.

  • Color coded according to membrane

    Fastest flow rate and lowest clogging of comparable alternatives

    0.1 µm PES available for mycoplasma protection

    0.2 µm PES devices are stem cell and mouse embryonic assay (MEA) tested

Ideal for the filtration of cell culture media, serum, additives and buffers.

Non-pyrogenic and endotoxin free - Rapid-Flow filter units meet the most rigorous testing standards for any vacuum filtration unit. Available with Fit for Purpose Sterile documentation.

Delivery: Comes assembled with 50 ml conical tube and a separate closure. Includes two reusable stands in each case.