Bottle top vacuum filtration systems, SFCA membrane, Rapid-Flow™, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Rapid-Flow™ disposable filter units feature the exclusive rapid-flow support column design, providing fast flow rates and high throughput.
Color coded according to membrane
- Fastest flow rate and lowest clogging of comparable alternatives
- 0.1 µm PES available for mycoplasma protection
- 0.2 µm PES devices are stem cell and mouse embryonic assay (MEA) tested
Ideal for the filtration of cell culture media, serum, additives and buffers.
Available with Fit for Purpose Sterile documentation
Non-pyrogenic and endotoxin free - Rapid-Flow filter units meet the most rigorous testing standards for any vacuum filtration unit.
Specifications
- Catalog No:
- 513-1621
- 513-1622
- 513-1611
- 734-5029
- 734-5070
- 513-1222
- 734-5069
- 734-5068
- 734-5067
- 513-1221
- 734-5066
- 734-5065
- 734-5064
- 734-5062
- 734-5063
- 513-1581
- 513-1571
- 513-1582
- 513-1572
- 513-1561
- 513-1551
- 513-1552
- 734-5061
- 513-1562
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)