ContactUsLinkComponent
Centrifuge filters, Vivaclear
Catalog # 516-0156
Supplier:  Sartorius
undefined
Centrifuge filters, Vivaclear
Catalog # 516-0156
Supplier:  Sartorius
Specifications
  • Pore size:
    0,80 µm
  • Pk:
    100
Specifications
About this item
Vivaclear centrifugal filters are disposable microfiltration devices for the fast and reliable clarification/filtration of biological samples in the range 100 to 500 µl. They can be used in fixed angle rotors suitable for 2,2 ml centrifuge tubes.
  • High-flux polyethersulfone membrane
  • Low hold-up volume (<5 µl)
  • Fast and reproducible performance
Applications: Clarification of samples before loading onto Vivapure protein purification spin columns; removal of particles and participates; filtration of plasma and serum; filtration of cells or cell debris.