Order Entry
Austria
ContactUsLinkComponent
 
Cable Box, 4-Way
Catalog # 139-1395
Supplier:  BACHMANN ELEKTRO
undefined
Cable Box, 4-Way
Catalog # 139-1395
Supplier:  BACHMANN ELEKTRO

Specifications

  • Description:
    Cable box, 4 Schuko sockets
  • Length:
    10 m
  • Pk:
    1

Specifications

About this item

Four-Way Schuko outlet with integrated one pole thermal cut-off switch.

  • Easy to roll and unroll
  • Cable (HO5VV-F G31.50 mm²) with double-earthed, right angle plug

Colour: White.

Max. power: Unrolled 3500 W; cable inside box 1200 W