Order Entry
Austria
Orders LinkContactUsLinkComponent
Staining dish, WHEATON®

Staining dish, WHEATON®

Staining dish, 80×55×35 mm
Catalog # 720-0701
Supplier:   DWK Life Sciences
Staining dish, WHEATON®
Catalog # 720-0701
Supplier:  DWK Life Sciences
Staining dish, WHEATON®
Staining dish, WHEATON®

Staining dish, 80×55×35 mm

Price
372,00 €
Log in for availability
1 * 6 ST
Product Details & Documents

Soda-lime glass.

  • Accommodates 19 slides arranged alternately straight across and diagonally

This staining dish, with lid, can hold 10 single standard slides of 76×26 mm or 20 slides back to back.

Specifications
  • Description:
    Staining dish
  • W×D×H:
    80×55×35 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.