Instrument trays, with lid
Supplier: BOCHEM
Certificates
About this item
Stainless steel, 18/10.
- Rounded corners for fast, easy cleaning
- Polished electrolytically, anti-magnetic
- Good chemical resistance, very good thermal resistance
Special lid with inset handle enabling the instrument trays to be stacked.
Frequently Bought Together