Instrument trays
Supplier: GLASWARENFABRIK KARL HECHT
Certificates
About this item
Glass with chrome-plated knobbed lid.
- Can be sterilised in hot air up to 180 °C
- Robust
- Good chemical resistance
Multipurpose dishes with vertical walls for a variety of applications in the laboratory, field use and industry.
Frequently Bought Together