Instrument dishes
Supplier: P.S. KUNSTSTOFWERKE
Certificates
About this item
Melamine dishes, white; PS lid, clear.
- Stable, good break-resistance
- Temperature resistant from −178 to +80 °C (melamine) and −40 to +55 °C (PS)
- Smooth surfaces for easy cleaning
Multipurpose dishes for a wide variety of laboratory applications. Also suitable for use in the food industry.
Frequently Bought Together