Orders LinkContactUsLinkComponent
Incinerating dishes | Avantor
Incinerating dishes
Incinerating dishes
Certificates
Incinerating dishes
Incinerating dishes
11 Options Available Below
Product Details & Documents
Porcelain dishes, glazed inside and out, except base.
  • Withstand maximum temperature 1000 °C
  • Shallow form without spout, deep form with spout
The rectangular dishes have sloping sides.
Measurements shown are W×D×H for rectangular dishes and Ø×H for others.
Product Family Options
Round dishes
Product Information
  • Type
    Capacity
    Ø ext.×H
    W×D×H
    Availability
    Price
  • Shallow
    8 ml
    37×10 mm
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,40 €
    9,40 €
    1 * 1 ST
       9,40 €
  • Shallow
    10 ml
    42×11 mm
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,70 €
    9,70 €
    1 * 1 ST
       9,70 €
  • Shallow
    15 ml
    48×12 mm
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,00 €
    13,00 €
    1 * 1 ST
       13,00 €
  • Deep
    15 ml
    40×21 mm
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,40 €
    9,40 €
    1 * 1 ST
       9,40 €
  • Deep
    30 ml
    50×25 mm
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,80 €
    11,80 €
    1 * 1 ST
       11,80 €
  • Deep
    60 ml
    60×31 mm
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,60 €
    14,60 €
    1 * 1 ST
       14,60 €
  • Flour (Mohs)
    30 ml
    56×23 mm
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,80 €
    11,80 €
    1 * 1 ST
       11,80 €
Rectangular dishes
Product Information
  • Type
    Capacity
    Ø ext.×H
    W×D×H
    Availability
    Price
  • Shallow
    10 ml
    -
    26×48×10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,80 €
    11,80 €
    1 * 1 ST
       11,80 €
  • Shallow
    20 ml
    -
    46×70×13 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,00 €
    20,00 €
    1 * 1 ST
       20,00 €
  • Shallow
    33 ml
    -
    42×55×16 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,30 €
    15,30 €
    1 * 1 ST
       15,30 €
  • Shallow
    55 ml
    -
    55×96×16 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,10 €
    27,10 €
    1 * 1 ST
       27,10 €
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.