Cell culture dishes, Nunc™
Supplier: Thermo Fisher Scientific
Certificates
About this item
PS (except 734-2113*, made of Permanox™), sterile, with lid. A large range of dishes, which have been tissue culture-treated.
- Optically clear and uniform surface suitable for microscopy
- 60 mm and 35 mm dishes available with grids for cloning or determination of plating efficiency
- Nunclon™Δ certified surface treatment
These cell culture/petri dishes are made of polystyrene.
These dishes receive the Thermo Scientific™ Nunclon™ Delta surface treatment, a proprietary surface modification that promotes maximum adhesion for a broad range of cell types.
Specifications
- Catalog No:
- 734-2042
- 734-2041
- 734-2043
- 734-2113
- 734-2704
- 734-2103
- 734-2114
- 734-1403
- 734-2045
- 734-2040
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement