Orders LinkContactUsLinkComponent
Desiccator plates
Desiccator plates
Supplier:   KARTELL
Desiccator plates
Supplier:  KARTELL
Certificates
Desiccator plates
Desiccator plates
3 Options Available Below
Product Details & Documents
Perforated PP desiccator plates. Suitable for use with desiccators: 467-2115, 467-2120 and 467-2125. Use at room temperature only, not suitable for use with hot crucibles.
Product Family Options
Product Information
  • Type
    Ø
    Availability
    Price
  • PP
    141 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,20 €
    11,20 €
    1 * 1 ST
       11,20 €
  • PP
    189 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,20 €
    16,20 €
    1 * 1 ST
       16,20 €
  • PP
    238 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,90 €
    25,90 €
    1 * 1 ST
       25,90 €