Desiccator cabinets, SICCO Star series
These transparent PMMA or borosilicate 3.3 glass cabinets have a reinforced aluminium frame, door with foam rubber gasket and magnetic strip. Designed for storing or drying humidity-sensitive products using silica gel. The controlled environment inside the desiccator is ideal for storing reference materials, retained samples, metallographic specimens, tobacco and DNA samples. Super-Star cabinets can be filled with inert gas, e.g. nitrogen to allow safe and secure storage of poisonous chemicals.
- Door mounted electronic hygrometer indicates minimum and maximum temperature and humidity values; hygrometers on Super-Star cabinets have a two-line display
- Temperature resistance from −20 to +70 °C
- Cabinets have non slip rubber feet and Star models are stackable
- Super-Star cabinets have a cylinder lock and include a hose with quick couplings 2×6 mm Ø gas inlet, non return valve and tubing connectors
: Supplied with four removable, acrylic glass, telescopic shelves, desiccant tray and silica gel. Capacity to accommodate a maximum of 26 shelves.
Frequently Bought Together