Stopcocks for dessicator
467-0078
- Description:Needle-valve stopcock, PTFE, cone size 24/29, aperture 0-4 mm
- Environmentally Preferable:
- Pk:1
Standard ground NS 24/29 Stopcocks, interchangeable, for tubulated cover of desiccators.
Standard ground NS 24/29 Stopcocks, interchangeable, for tubulated cover of desiccators.