Orders LinkContactUsLinkComponent
Micro Cells for Absorption Measurements
Micro Cells for Absorption Measurements
Supplier:   Hellma
Micro Cells for Absorption Measurements
Supplier:  Hellma
Certificates
Micro Cells for Absorption Measurements
Micro Cells for Absorption Measurements
10 Options Available Below
Product Details & Documents
Hellma® high precision macro cells from quartz glass or special optical glass for absorption measurements in the Vis range, 10 mm path length, with lid.
  • Cells from quartz glass or special optical glass
  • Wavelength range from 200 to 2500 nm
  • Path length: 10 mm
  • PTFE lid or stopper
  • Volume: 300 to 700 µl
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers. Highest precision: Hellma® precision cells are characterised by the highest precision in path length accuracy and parallelism. This guarantees continuously reproducible measurement results.
Chemically resistant and thermally bonded: Hellma® precision cells are thermally bonded and not glued, which gives them a very high resistance to temperatures and chemicals. This prevents unintentional leakage of liquids. Excellent transmission: By using selected high-performance quartz glasses, Hellma® precision cells achieve excellent transmission >82% for exact measurement results.
Secure handling: Hellma® precision cells have chamfered edges that enable safe and comfortable handling. In addition, the risk of splintering is minimised. Sustainable: Hellma® precision cells can be easily cleaned and reused and thus remain in use for a long time. A sustainable and environmentally friendly alternative.
Certifications: Hellma® precision cells are manufactured according to EN ISO 9001/EN ISO 14001 and meet the highest optical standards. The declaration of compliance is available for download on the Hellma website. Further certificates (factory certificate 2.2 and acceptance test certificate 3.1) are available on request.
Ordering information: Cell pairs are spectrally calibrated.
Documents
Product Family Options
Micro cells with PTFE stoppers
Product Information
  • Type
    Ext.Ø
    Height
    Centre height
    Volume
    Path length
    Window material
    Availability
    Price
  • 634-9242
    With PTFE stopper
    With PTFE stopper
    Micro
    12,5×12,5 mm
    40 mm
    8,5 - 20 mm
    400 µl
    10 mm
    QS: Quartz glass, high performance for measurements in the UV/Vis range from 200 - 2500 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 202,00 €
    202,00 €
    1 * 1 ST
       202,00 €
  • 634-9241
    With PTFE stopper, black side walls and base
    With PTFE stopper, black side walls and base
    Micro
    12,5×12,5 mm
    40 mm
    8,5 - 20 mm
    400 µl
    10 mm
    QS: Quartz glass, high performance for measurements in the UV/Vis range from 200 - 2500 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 380,00 €
    380,00 €
    1 * 1 ST
       380,00 €
Micro cell with PTFE lids
Product Information
  • Type
    Ext.Ø
    Height
    Centre height
    Volume
    Path length
    Window material
    Availability
    Price
  • 634-9236
    With PTFE lid
    With PTFE lid
    Micro
    12,5×12,5 mm
    25 mm
    8,5 - 15 mm
    300 µl
    10 mm
    QS: Quartz glass, high performance for measurements in the UV/Vis range from 200 - 2500 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 150,00 €
    150,00 €
    1 * 1 ST
       150,00 €
  • 634-9240
    With PTFE lid
    With PTFE lid
    Micro
    12,5×12,5 mm
    45 mm
    15 - 20 mm
    500 µl
    10 mm
    QS: Quartz glass, high performance for measurements in the UV/Vis range from 200 - 2500 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 195,00 €
    195,00 €
    1 * 1 ST
       195,00 €
  • 634-1060
    With PTFE lid, black side walls and base
    With PTFE lid, black side walls and base
    Micro
    12,5×12,5 mm
    45 mm
    15 - 20 mm
    500 µl
    10 mm
    QS: Quartz glass, high performance for measurements in the UV/Vis range from 200 - 2500 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 369,00 €
    369,00 €
    1 * 1 ST
       369,00 €
  • HELL104-002-10-40
    With PTFE lid
    With PTFE lid
    Micro
    12,5×12,5 mm
    45 mm
    8,5 - 20 mm
    700 µl
    10 mm
    QS: Quartz glass, high performance for measurements in the UV/Vis range from 200 - 2500 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
    Specifications:
    New Product
    New Product
  • 634-9080
    With PTFE lid
    With PTFE lid
    Micro
    12,5×12,5 mm
    45 mm
    8,5 - 20 mm
    700 µl
    10 mm
    QS: Quartz glass, high performance for measurements in the UV/Vis range from 200 - 2500 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 169,00 €
    169,00 €
    1 * 1 ST
       169,00 €
  • 634-9079
    With PTFE lid
    With PTFE lid
    Micro
    12,5×12,5 mm
    45 mm
    8,5 - 20 mm
    700 µl
    10 mm
    Special optical glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,30 €
    86,30 €
    1 * 1 ST
       86,30 €
  • 634-9081
    With PTFE lid, black side walls and base
    With PTFE lid, black side walls and base
    Micro
    12,5×12,5 mm
    45 mm
    8,5 - 20 mm
    700 µl
    10 mm
    QS: Quartz glass, high performance for measurements in the UV/Vis range from 200 - 2500 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 279,00 €
    279,00 €
    1 * 1 ST
       279,00 €
Micro cells with PTFE lids
Product Information
  • Type
    Ext.Ø
    Height
    Centre height
    Volume
    Path length
    Window material
    Availability
    Price
  • 634-9214
    Micro cell with PTFE lid and black windows
    Micro cell with PTFE lid and black windows
    Micro
    45 mm
    700 µl
    10 mm
    Optical glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 132,00 €
    132,00 €
    1 * 1 ST
       132,00 €