Orders LinkContactUsLinkComponent
Faeces containers
Faeces containers
Supplier:   KARTELL
Faeces containers
Supplier:  KARTELL
Certificates
Faeces containers
Faeces containers
3 Options Available Below
Product Details & Documents
PS, transparent. Non sterile or sterile.
  • With PP spatula incorporated into the cap
Documents
Product Family Options
Product Information
  • Capacity
    Sterility
    Ø
    Height
    Availability
    Price
  • 15 ml
    Non sterile
    22 mm
    63 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 246,00 €
    246,00 €
    1 * 1.000 ST
       246,00 €
  • 60 ml
    Non sterile
    41 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 360 ST
    1 * 360 ST - 195,00 €
    195,00 €
    1 * 360 ST
       195,00 €
  • 60 ml
    Sterile
    41 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 359,00 €
    359,00 €
    1 * 400 ST
       359,00 €