Containers, straight, wide mouth, with screw cap, Corning® Gosselin™
Supplier: Corning
Certificates
About this item
PP translucent or PS clear, with PE screw cap. Straight walled vessels for sample collection, storage and transport. Twist closure for quick opening and closing, with leakproof inner lip. PS offers good thermal and chemical resistance. PP offers excellent thermal and chemical resistance.
- Selection of colour coded closures for easier sample identification
- Available sterile or packed under aseptic conditions
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Catalog No:
- 216-2094
- 216-2093
- GOSSTP30C-003
- 216-2095
- GOSSTP53C-042
- 216-2104
- GOSSTP53C-032
- 216-2089
- 216-2092
- 216-2091
- GOSSTP53C-052
- 216-2088
- GOSSTP53C-030
- 216-2102
- 215-1324
- 216-2113
- GOSSTP30C-004
- 216-2111
- 216-2100
- 216-2086
- 216-2099
- GOSSTP30C-032
- 216-2087
- GOSSTP53C-014
- GOSSTP175-06
- GOSSTP30C-014
- 216-2085
- GOSSTP30C-021
- 216-2096
- 216-2098
- 216-2097
- 216-2084
- 216-0518
- GOSSTP53C-023
- GOSSTP53C-021
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre