Orders LinkContactUsLinkComponent
Sample containers, with snap-on lids
Sample containers, with snap-on lids
Supplier:   KARTELL
Sample containers, with snap-on lids
Supplier:  KARTELL
Certificates
Sample containers, with snap-on lids
Sample containers, with snap-on lids
8 Options Available Below
Product Details & Documents
LDPE, translucent.
  • Good chemical resistance
  • Heat resistant up to 80 °C
  • Light and sturdy design
With attached snap-on lid. The lid snaps easily into place with light pressure. Flat base.
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Availability
    Price
  • 1,0 ml
    8,0 mm
    32,0 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 186,00 €
    186,00 €
    1 * 500 ST
       186,00 €
  • 2,5 ml
    14,0 mm
    31,5 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 35,30 €
    35,30 €
    1 * 100 ST
       35,30 €
  • 8,0 ml
    17,2 mm
    57,6 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 58,00 €
    58,00 €
    1 * 100 ST
       58,00 €
  • 7,0 ml
    22,8 mm
    32,9 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 104,00 €
    104,00 €
    1 * 100 ST
       104,00 €
  • 20,0 ml
    24,8 mm
    74,5 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 79,60 €
    79,60 €
    1 * 100 ST
       79,60 €
  • 35,0 ml
    31,0 mm
    74,5 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 53,30 €
    53,30 €
    1 * 50 ST
       53,30 €
  • 25,0 ml
    31,3 mm
    52,5 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 49,60 €
    49,60 €
    1 * 50 ST
       49,60 €
  • 5,0 ml
    15,0 mm
    49,5 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 51,30 €
    51,30 €
    1 * 100 ST
       51,30 €