Orders LinkContactUsLinkComponent
Sample containers, with snap-on caps, Nalgene®
Sample containers, with snap-on caps, Nalgene®
   Sustainable Options Available
Sample containers, with snap-on caps, Nalgene®
   Sustainable Options Available
Certificates
Sample containers, with snap-on caps, Nalgene®
Sample containers, with snap-on caps, Nalgene®
6 Options Available Below
Product Details & Documents
LDPE, translucent.
  • Good chemical resistance
  • Heat resistant from –100 to +80 °C
  • Tight fitting snap-on cover, robust
Suitable as a sample container, beaker for pH measurements, weighing container and many other applications.
Specifications for Product Family
Specifications
  • Catalog No:
  • 216-8151
  • 216-8152
  • 216-8153
  • 216-8154
  • 216-8155
  • 216-8156
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Availability
    Price
  • 5 ml
    23,6 mm
    24,9 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 14,70 €
    14,70 €
    1 * 12 ST
       14,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8151
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 12 ml
    27,1 mm
    35,6 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 16,40 €
    16,40 €
    1 * 12 ST
       16,40 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8152
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 18 ml
    27,1 mm
    52,6 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 22,50 €
    22,50 €
    1 * 12 ST
       22,50 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8153
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 28 ml
    27,1 mm
    84,6 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 28,00 €
    28,00 €
    1 * 12 ST
       28,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8154
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 50 ml
    37,3 mm
    70,9 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 35,40 €
    35,40 €
    1 * 12 ST
       35,40 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8155
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 75 ml
    37,3 mm
    107,3 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 47,90 €
    47,90 €
    1 * 12 ST
       47,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8156
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact