Jars, wide mouth, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PC, clear with white PP screw cap, PMP, clear with white PP screw cap or PPCO, translucent with white PP screw cap.
- PC: Extremely shock-proof, non toxic. Suitable for applications in cold rooms and refrigerators. Temperature resistant from −135 to +135 °C
- PMP: Good chemical resistance, ideal as a storage container; formaldehyde loss only 2% per year. Complies with the requirements of CFR 21, Part 177.1520 for use with food and drink. Temperature resistant from −20 to +175 °C
- PPCO: Very good chemical resistance. Temperature resistant from −10 to +121 °C
Straight-sided containers with wide opening.
Specifications
- Catalog No:
- 216-8214
- 216-8215
- 216-8207
- 216-8206
- 216-8221
- 216-8213
- 216-8225
- 216-8226
- 216-8227
- 216-8205
- 216-8203
- 216-8224
- 216-8204
- 216-8201
- 216-8222
- 216-8202
- 216-8223
- 216-8216
- 216-8217
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)