Adapters with tube connector
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Adapter with standard ground joint cone and straight or right angled tube connector. The connector has an external diameter of 8 mm.
Frequently Bought Together