Coil condensers with standard ground joints
DURAN®, borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
For cooling condensate from vapours, with cone and socket. Water supplied through plastic connectors with 8 mm external diameter and GL 14 threaded joints.
Frequently Bought Together