Allihn condensers with standard ground joints
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With core and jacket. Water supplied through plastic connectors with 8 mm external diameter and GL 14 threaded joints.
201-1168 and 201-1678 according to DIN 12581
Certifications: 201-1168 and 201-1678 according to DIN 12581
Frequently Bought Together