Condenser with cooling coil and jacket, with standard ground joints
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
DURAN®, borosilicate 3.3 glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Additional cooling surface for condensing highly volatile vapours. Cooling coil and cooling jacket, with standard ground cone and socket. Water supplied through plastic connectors with 8 mm external diameter and GL 14 threaded joints.
DIN 12593
Certifications: DIN 12593
Frequently Bought Together