A fast, versatile HPLC column packing material. These columns rapidly and easily separate closely related alcohols and organic acids without column heating. The sulfonated poly(styrene-divinylbenzene) support separates samples by a mixed mode mechanism. A wide range of samples can be analysed with these columns because the selectivity of the column can be altered by changing the pH of the buffer or adding an organic modifier (methanol, acetonitrile) to the buffer. The stability to organic solvents makes it possible to analyse samples that have been too highly retained on conventional ion exclusion supports.
- Use with organic solvent from 0 to 100% for elution of highly retained compounds
The excellent selectivity of the PRP-X300 columns ensures most separations are complete within five minutes. The high performance packing ensures narrow peaks under isocratic conditions. The use of isocratic conditions allows samples to be analyzed without column reequilibration. When samples are too complex or the isocratic analysis takes too long, you can always run a gradient. Most samples require only minimal preparation before injection.
Compound classification: Organic acids/Alcohols
- Catalog No:
- HAMI79915
- 554-1249
- 554-0255
- 554-1229
- 554-1250
- HAMI79289
- 554-1274
- 554-0247
- 554-1268
- 554-0351
- 554-1230
- Exchange capacity:
- 0,17 meq/gm
- 0,17 meq/gm
- 0,17 meq/gm
- 0,17 meq/gm
- 0,17 meq/gm
- 0,17 meq/gm
- 0,17 meq/gm
- 0,17 meq/gm
- 0,17 meq/gm
- 0,17 meq/gm
- 0,17 meq/gm
- Max. pressure:
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- pH range:
- 1-13
- 1-13
- 1-13
- 1-13
- 1-13
- 1-13
- 1-13
- 1-13
- 1-13
- 1-13
- 1-13
- Temperature limit:
- 5-60 °C
- 5-60 °C
- 5-60 °C
- 5-60 °C
- 5-60 °C
- 5-60 °C
- 5-60 °C
- 5-60 °C
- 5-60 °C
- 5-60 °C
- 5-60 °C
- USP code:
- L22
- L22
- L22
- L22
- L22
- L22
- L22
- L22
- L22
- L22
- L22