Orders LinkContactUsLinkComponent
End plug
End plug
End plug for 1/4 28 ports
Catalog # 554-1064
Supplier:   VICI JOUR RESEARCH
End plug
Catalog # 554-1064
Supplier:  VICI JOUR RESEARCH
End plug
End plug
End plug for 1/4 28 ports
Price
66,70 €
Log in for availability
1 * 5 ST
Product Details & Documents
Plugs can be used to close off an unused port in a valve or manifold. Size ¹/₄ - 28".
Specifications
  • Description:
    End plug for 1/4 28 ports