Orders LinkContactUsLinkComponent
VWR® Hichrom Precolumn Filters
VWR® Hichrom Precolumn Filters
Supplier:   Avantor
VWR® Hichrom Precolumn Filters
Supplier:  Avantor
Certificates
VWR® Hichrom Precolumn Filters
VWR® Hichrom Precolumn Filters
8 Options Available Below
Product Details & Documents
Placed immediately before the column, the precolumn filter traps particles introduced with the sample. For samples, which may irreversibly absorb onto the column, the use of guard cartridges is strongly recommended.
  • Protect columns from particulates, ultra low dead volume
  • Virtually no dead volume with no band broadening or loss of efficiency
  • Compatible with all HPLC columns
Filter HI-704 comprises a stainless steel holder with a 0,020" thru hole and a 2 µm PEEK encapsulated stainless steel frit (supplied).
The fingertight HI-685 precolumn filter includes a 2 µm stainless steel frit which is contained within a PEEK fitting connecting directly onto the column.
Product Family Options
Product Information
  • For column
    Porosity
    Availability
    Price
  • HI-685
    ColumnSaver fingertight precolumn filters
    ColumnSaver fingertight precolumn filters
    5 μm
    2,0 µm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 383,00 €
    383,00 €
    1 * 10 ST
       383,00 €
  • HI-686
    ColumnSaver fingertight precolumn filters
    ColumnSaver fingertight precolumn filters
    3 μm
    0,5 µm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 383,00 €
    383,00 €
    1 * 10 ST
       383,00 €
  • 554-7722
    HI-704, precolumn filter assembly (complete with frit)
    HI-704, precolumn filter assembly (complete with frit)
    2,0 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 140,00 €
    140,00 €
    1 * 1 ST
       140,00 €
  • HI-615
    Precolumn filter
    Precolumn filter
    0,5 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 182,00 €
    182,00 €
    1 * 1 ST
       182,00 €
  • HICHHI-602
    Precolumn filter
    Precolumn filter
    UHPLC
    0,5 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 92,10 €
    92,10 €
    1 * 1 ST
       92,10 €
  • HICHHI-602X
    Precolumn filters
    Precolumn filters
    UHPLC
    0,5 µm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 785,00 €
    785,00 €
    1 * 10 ST
       785,00 €
Replacement frits
Product Information
  • For column
    Porosity
    Availability
    Price
  • 554-7720
    HI-101, PEEK encapsulated stainless steel frit
    HI-101, PEEK encapsulated stainless steel frit
    2,0 µm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 81,30 €
    81,30 €
    1 * 10 ST
       81,30 €
  • 554-7719
    HI-102, PEEK encapsulated stainless steel frit
    HI-102, PEEK encapsulated stainless steel frit
    0,5 µm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 90,90 €
    90,90 €
    1 * 10 ST
       90,90 €