Orders LinkContactUsLinkComponent
ValvTool
ValvTool
ValvTool, 1/4" and 5/16" open wrench, 1/4" hex and 5/16" heads
Catalog # 554-1108
Supplier:   VICI JOUR RESEARCH
ValvTool
Catalog # 554-1108
Supplier:  VICI JOUR RESEARCH
ValvTool
ValvTool
ValvTool, 1/4" and 5/16" open wrench, 1/4" hex and 5/16" heads
Price
72,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
For opening and tightening ¹/₄'' nuts.
  • Slotted head allows tightening with tubing in situ
Specifications
  • Description:
    ValvTool, 1/4" and 5/16" open wrench
  • Use with head style:
    1/4" hex and 5/16" heads