Nuts with double ferrules
About this item
These VICI Jour® nuts incorporate a unique PEEK double ferrule which grips the tubing in two places for added security, especially with PEEK or Tefzel tubing. For virtually any ¹/₁₆'' OD fittings eg. Parker, Swagelok, Waters, Valco, Rheodyne, Upchurch.
- Inert all-PEEK construction
- Fingertight and hex-head versions
- Rated up to 420 bar (6000 psi)
5 Options Available Below
Product Details & Documents
These VICI Jour® nuts incorporate a unique PEEK double ferrule which grips the tubing in two places for added security, especially with PEEK or Tefzel tubing. For virtually any ¹/₁₆'' OD fittings eg. Parker, Swagelok, Waters, Valco, Rheodyne, Upchurch.
- Inert all-PEEK construction
- Fingertight and hex-head versions
- Rated up to 420 bar (6000 psi)
Product Family Options
Product Information
- AvailabilityPrice
- JOURJR-5510-5Hex-head nut, long
- JOURJR-5511-5Hex-head nut, short
- JOURJR-5580-5Fingertight nut
- 554-1067Double ferrule
- JOURJR-5004-5Double ferrule