Accessories for Chromolith®
Supplier: Merck
Certificates
About this item
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
EPA screw neck vials, N 24
Supplier: MACHEREY-NAGEL
Volumetric flasks, class A, blue markings
Supplier: HIRSCHMANN
VWR®, Fire Blankets
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Hichrom Column End Plugs
Supplier: Avantor
Biocompatible precolumn filters, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Aminophenol, Reference Standards
Supplier: USP
Precision thermometers
Supplier: Amarell