Orders LinkContactUsLinkComponent
HPLC columns, preparative scale, Chromolith® Prep
HPLC columns, preparative scale, Chromolith® Prep
Supplier:   Merck
HPLC columns, preparative scale, Chromolith® Prep
Supplier:  Merck
Certificates
HPLC columns, preparative scale, Chromolith® Prep
HPLC columns, preparative scale, Chromolith® Prep
5 Options Available Below
Product Details & Documents
Chromolith® Prep monolithic stationary phases are new ultrapure silica phases which gain their special properties from a bimodal pore structure. This structure is a property of the new 'Sol-Gel' technology and consists of macropores and mesopores. The mesopores with an average diameter of 12 nm, form the fine porous structure of the column interior and create a very large surface area on which adsorption of the target compounds occurs.
The large macropores, with a pore diameter ranging from 3 µm, form a dense network of pores and allow a high flow rate due to a low resistance factor. The resulting excellent accessibility of the mesopores (total porosity >80%) ensures fast adsorption and desorption kinetics because of the short diffusion length inside the pores. This results in dramatically reduced separation times and thus greatly increased productivity.
To operate a monolithic column it is necessary to ensure that no flow through occurs in the wall region. Therefore the monolith is cladded with a polymeric material (special PEEK) and can be connected directly to each HPLC system and used as 'ready to use' column.
Product Family Options
Semi preparative
Product Information
  • Column Format
    Phase
    Ø int.
    Length
    Availability
    Price
  • 1.52015.0001
    Chromolith® SemiPrep Si
    Chromolith® SemiPrep Si
    Semi-preparative
    Si
    10,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 890,00 €
    1 890,00 €
    1 * 1 ST
       1 890,00 €
  • 1.52016.0001
    Chromolith® SemiPrep RP-18 endcapped
    Chromolith® SemiPrep RP-18 endcapped
    Semi-preparative
    RP-18e
    10,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 330,00 €
    2 330,00 €
    1 * 1 ST
       2 330,00 €
Guard cartridges
Product Information
  • Column Format
    Phase
    Ø int.
    Length
    Availability
    Price
  • 1.52011.0001
    Chromolith® Si, guard column
    Chromolith® Si, guard column
    Guard cartridge
    Si
    4,60 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 282,00 €
    282,00 €
    1 * 3 ST
       282,00 €
  • 1.52036.0001
    Chromolith® SemiPrep RP-18 endcapped, guard column
    Chromolith® SemiPrep RP-18 endcapped, guard column
    Guard cartridge
    RP-18e
    10,00 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 740,00 €
    740,00 €
    1 * 1 ST
       740,00 €
Preparative
Product Information
  • Column Format
    Phase
    Ø int.
    Length
    Availability
    Price
  • 1.25251.0001
    Chromolith® Prep Si
    Chromolith® Prep Si
    Preparative
    Prep Si
    25,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Accessories
Product Information
  • Catalog #
    Ø int.
    Length
    Availability
    Price
  • Chromolith® SemiPrep guard cartridge holder
    Chromolith® SemiPrep guard cartridge holder
    1.52037.0001
    10,00 mm
    10 mm
    Log in for availability
    1 * 1 ST - 630,00 €