Orders LinkContactUsLinkComponent
HPLC columns, LiChrosorb®
HPLC columns, LiChrosorb®
Supplier:   Merck
HPLC columns, LiChrosorb®
Supplier:  Merck
Certificates
HPLC columns, LiChrosorb®
17 Options Available Below
Product Details & Documents
LiChrosorb® is one of the most successful HPLC packing materials. It is a totally porous, irregular particle silica gel produced with pore sizes of 60 and 100 Å. LiChrosorb® HPLC columns are available either as a cartridge, requiring re-usable endfittings, or as a complete end-fitted column.
If guard columns are required, use LiChrospher® guard cartridges listed in LiChrospher® section.
Product Family Options
LiChrosorb® Hibar® columns with fixed endfittings
Product Information
  • Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • RP-18
    100 Å
    5 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 803,00 €
    803,00 €
    1 * 1 ST
       803,00 €
  • RP-18
    100 Å
    5 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 921,00 €
    921,00 €
    1 * 1 ST
       921,00 €
  • RP-18
    100 Å
    10 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 875,00 €
    875,00 €
    1 * 1 ST
       875,00 €
  • RP-18
    100 Å
    5 µm
    4,60 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 921,00 €
    921,00 €
    1 * 1 ST
       921,00 €
  • RP-8
    100 Å
    5 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 767,00 €
    767,00 €
    1 * 1 ST
       767,00 €
  • RP-8
    100 Å
    5 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 921,00 €
    921,00 €
    1 * 1 ST
       921,00 €
  • RP-8
    100 Å
    10 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 875,00 €
    875,00 €
    1 * 1 ST
       875,00 €
  • RP-8
    100 Å
    5 µm
    4,60 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 921,00 €
    921,00 €
    1 * 1 ST
       921,00 €
  • Si
    60 Å
    5 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 875,00 €
    875,00 €
    1 * 1 ST
       875,00 €
LiChroCART® 4 mm ID cartridge
Product Information
  • Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • RP-18
    100 Å
    5 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 689,00 €
    689,00 €
    1 * 1 ST
       689,00 €
  • RP-18
    100 Å
    5 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 828,00 €
    828,00 €
    1 * 1 ST
       828,00 €
  • RP-18
    100 Å
    10 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 786,00 €
    786,00 €
    1 * 1 ST
       786,00 €
  • RP-8
    100 Å
    5 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 689,00 €
    689,00 €
    1 * 1 ST
       689,00 €
  • RP-8
    100 Å
    5 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 828,00 €
    828,00 €
    1 * 1 ST
       828,00 €
  • RP-8
    100 Å
    10 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 786,00 €
    786,00 €
    1 * 1 ST
       786,00 €
  • Si
    60 Å
    5 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 656,00 €
    656,00 €
    1 * 1 ST
       656,00 €
  • Si
    60 Å
    5 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 824,00 €
    824,00 €
    1 * 1 ST
       824,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Guard cartridge holder, stand-alone, for capillary connection of guard columns LiChroCart® 4-4 to column Hibar® RT
    Guard cartridge holder, stand-alone, for capillary connection of guard columns LiChroCart® 4-4 to column Hibar® RT
    1.16217.0001
    Log in for availability
    1 * 1 ST - 296,00 €