Orders LinkContactUsLinkComponent
HPLC columns, Purospher®
HPLC columns, Purospher®
Supplier:   Merck
HPLC columns, Purospher®
Supplier:  Merck
Certificates
HPLC columns, Purospher®
HPLC columns, Purospher®
9 Options Available Below
Product Details & Documents
Purospher® RP-18 is designed for the separation of problematic basic compounds with simple neutral eluents. In addition Purospher® RP-18 allows the separation of hydrophilic compounds with aqueous mobile phase. Purospher® RP-18 is based upon a high purity, metal-free silica. The multi-step chemical modification and deactivation of the surface, and the polar endcapping, enables a peak-tailing-free elution of basic and chelating compounds. Purospher® RP-18 is not suitable for the separation of acidic compounds. The good chemical stability of Purospher® RP-18 enables the use of mobile phase conditions at pH 8 for a long period of time.
Purospher® RP-18e is a versatile, high purity HPLC column producing excellent peak shapes, designed for both the separation of basic compounds with simple neutral eluents, and for the elution of strongly acidic compounds. Excellent separations in a shorter period of time with very good peak symmetry saves time and money. The excellent balance of Purospher® RP-18e, in the sum of its chromatographic properties, is the key to better separations of complex samples with simpler, neutral eluents. The good chemical stability of Purospher® RP-18e enables the use of mobile phase conditions at pH 8 for a long period of time.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a 4×4 mm guard column.
Product Family Options
Purospher® RP-18
Product Information
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • Guard column
    RP-18
    90 Å
    5 µm
    4,00 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 580,00 €
    580,00 €
    1 * 10 ST
       580,00 €
  • Analytical
    RP-18
    90 Å
    5 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 040,00 €
    1 040,00 €
    1 * 1 ST
       1 040,00 €
  • Analytical
    RP-18
    90 Å
    5 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 170,00 €
    1 170,00 €
    1 * 1 ST
       1 170,00 €
Purospher® RP-18e
Product Information
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • Guard column
    RP-18e
    90 Å
    5 µm
    4,00 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 696,00 €
    696,00 €
    1 * 10 ST
       696,00 €
  • Analytical
    RP-18e
    90 Å
    5 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 100,00 €
    1 100,00 €
    1 * 1 ST
       1 100,00 €
  • Analytical
    RP-18e
    90 Å
    5 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 170,00 €
    1 170,00 €
    1 * 1 ST
       1 170,00 €
  • Analytical
    RP-18e
    90 Å
    5 µm
    3,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 040,00 €
    1 040,00 €
    1 * 1 ST
       1 040,00 €
  • Analytical
    RP-18e
    90 Å
    5 µm
    3,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 2 190,00 €
    2 190,00 €
    1 * 3 ST
       2 190,00 €
  • Analytical
    RP-18e
    90 Å
    5 µm
    3,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 020,00 €
    1 020,00 €
    1 * 1 ST
       1 020,00 €