Orders LinkContactUsLinkComponent
Chiral columns, ChiraDex® | Avantor
Chiral columns, ChiraDex®
Supplier:   Merck
Chiral columns, ChiraDex®
Supplier:  Merck
Certificates
Chiral columns, ChiraDex®
3 Options Available Below
Product Details & Documents
ChiraDex® is a versatile HPLC column characterised by broad enantioselectivity and can be used for the separation of enantiomers of numerous different classes of substances. ChiraDex® is based on beta-cyclodextrin covalently linked to spherical particles of silica, and is ideal for chiral separation of hydrocarbons, steroids, phenol esters and derivatives, aromatic amines, heterocycles with five- to seven-membered ring. Simply composed RP-eluents can be used in most separations.
The LiChroCART® cartridges require the use of the manu-CART® NT cartridge holder 1.51486.0001.
Dokumente
Specifications for Product Family
Specifications
  • Catalog No:
  • 1.50117.0001
  • 1.51333.0001
  • 1.51000.0001
  • pH range:
  • 3-7,5
  • 3-7,5
  • 3-7,5
  • Surface area:
  • 300-360 m²/g
  • 300-360 m²/g
  • 300-360 m²/g
Specifications
Product Family Options
Product Information
  • Column Format
    Column hardware
    Pore size
    Particle Size
    Ø int.
    Length
    Packing material
    Availability
    Price
  • ChiraDex®
    Guard column
    Stainless steel
    100 Å
    5 µm
    4,00 mm
    4 mm
    Spherical silica
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 542,00 €
    542,00 €
    1 * 10 ST
       542,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.50117.0001
    pH range
    3-7,5
    Surface area
    300-360 m²/g
  • ChiraDex®
    Analytical
    Stainless steel
    100 Å
    5 µm
    4,00 mm
    250 mm
    Spherical silica
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 680,00 €
    1 680,00 €
    1 * 1 ST
       1 680,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.51333.0001
    pH range
    3-7,5
    Surface area
    300-360 m²/g
  • 1.51000.0001
    ChiraDex® High Resolution
    ChiraDex® High Resolution
    Analytical
    Stainless steel
    100 Å
    5 µm
    4,00 mm
    250 mm
    Spherical silica
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 1 680,00 €
    1 680,00 €
    1 * 1 KIT
       1 680,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.51000.0001
    pH range
    3-7,5
    Surface area
    300-360 m²/g