ContactUsLinkComponent
Sleeve protectors, Secutex® pro, PP/PE
Sleeve protectors, Secutex® pro, PP/PE
Catalog # ZVGA30241-90
undefined
Sleeve protectors, Secutex® pro, PP/PE
Catalog # ZVGA30241-90
Specifications
  • Garment application:
    Laboratory
  • Gender:
  • Colour:
    White
  • Length:
    380 mm
  • Environmentally Preferable:
  • Pk:
    100 PAIR
Specifications
About this item
These laminate sleeve protectors provide protection against fine dust and aggressive and toxic substances. Fitted with two elasticated cuffs.
  • Lint-free, resistant to abrasion, tear and water
  • Permeable to water vapour which ensures breathability
  • Anti-static