ContactUsLinkComponent
Laboratory overalls, SMS
Laboratory overalls, SMS
Catalog # 113-0365
undefined
Laboratory overalls, SMS
Catalog # 113-0365
Specifications
  • Clothing Size:
    L
  • Garment application:
    Laboratory,
    Food
  • Colour:
    White
  • Seq:
    03
  • Pk:
    25
  • Weight:
    60 g
Specifications
About this item
Overalls in different types and colours are made of spunbond/meltblown/spunbond material.
  • Soft, breathable material for high mobility and comfort
  • Provides splash protection and particulate barrier
Ideal for industrial, pharmaceutical, food processing, construction sites, and certain hazardous and non hazardous sites.
Weight: 60 g
Colour: White or blue