ContactUsLinkComponent
Laboratory coats, Kimtech™ A7, disposable
Laboratory coats, Kimtech™ A7, disposable
Catalog # 113-1005
Supplier:  KIMBERLY CLARK
undefined
Laboratory coats, Kimtech™ A7, disposable
Catalog # 113-1005
Supplier:  KIMBERLY CLARK
Specifications
  • Clothing Size:
    L
  • Closure type:
    Press studs / snaps
  • Garment application:
    Chemical protection,
    Laboratory
  • Gender:
    Unisex
  • Pockets:
    With pockets
  • Colour:
    White
  • Seq:
    03
  • Pk:
    15
Specifications
About this item
These anti-static lab coats are constructed of multiple layers, with a chemical and abrasion resistant spunbond 100% PP outer layer, and microfibre, cloth-like inner layer designed to filter out a variety of water-based liquids and dry particulates.
  • Three fabric layers for enhanced protection
  • Triple over-locked seams for strength, durability and security, even with heavy use
  • Breathable material decreases the risk of heat stress to enable long periods of uninterrupted wear
  • Excellent particle and bacterial filtration
  • Tear resistant construction, surface resistivity limitation
  • Latex-free and silicone-free, low linting
  • Type 6 limited chemical splash protection and rigorously tested against an array of contaminants
Designed to protect analysts from low risk liquid chemicals in the form of sprays, aerosols and light splashes (type 6 protection).
Colour: white
Certifications: Type 6 (PB); Cat. III