ContactUsLinkComponent
Aprons, Secutex® pro, Z 40, PP/PE
Aprons, Secutex® pro, Z 40, PP/PE
Catalog # 114-3441
undefined
Aprons, Secutex® pro, Z 40, PP/PE
Catalog # 114-3441
Specifications
  • Garment application:
    Chemical protection
  • Gender:
  • Colour:
    White
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
This disposable apron made from laminated anti-static PP/PE protects against aggressive and toxic substances.
  • Lint-free, water and particle-proof
  • Long version with neckband and belt
Colour: white