Orders LinkContactUsLinkComponent
Threaded closures for rectangular bottles
Threaded closures for rectangular bottles
Screw cap, PP 28, White
Catalog # MUEL14.063.01
Supplier:   MUELLER KREMPEL
Threaded closures for rectangular bottles
Catalog # MUEL14.063.01
Supplier:  MUELLER KREMPEL
Threaded closures for rectangular bottles
Screw cap, PP 28, White
Price
14,60 €
Log in for availability
1 * 20 ST
Product Details & Documents
PP, white.
  • Tamper-evident closure
  • Self-sealing with internal cone
  • Can only be used for rectangular bottles 215-8331 to 215-8334
Specifications
  • Colour:
    White
  • Description:
    Screw cap, PP 28