Tubing clamps, polyester, Keck™, DURAN®
Lieferant: DWK Life Sciences
Zertifikate
Über diesen Artikel
Polyester (PBT). For smooth control of flow rates in tube connections.
- Very good thermal resistance (operating temperature up to +180 °C)
- Autoclavable
- Convenient, easy operation, colour coded according to size
Frequently Bought Together