Fork clamps for spherical joints
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
Clips for spherical joints made from stainless steel chrome/nickel.
- Easy to install and remove
- Excellent heat and chemical resistance
- With set screw
Frequently Bought Together