Joint clamps for conical ground joints
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
POM, coloured. For firm, securely fitting conical ground joints. These clamps are made of plastic to avoid scratching the glass, as happens with conventional metal joint clamps.
- Very good chemical resistance
- Heat resistant up to +150 °C
- Sturdy, easy to install and remove
Frequently Bought Together