Clips for standard ground joints
Nickel-chromium steel. For conical ground joint connections.
- Quick, easy assembly and disassembly
- Very good thermal resistance, good chemical resistance
: Optional fixing screws are available.
Frequently Bought Together