VWR®, Clips for Conical Standard Ground Joints
PTFE with inner steel spring. Very stable, safe and durable, easy to use.
- Excellent chemical resistance
- Temperature resistant from −200 to +280 °C
Frequently Bought Together
PTFE with inner steel spring. Very stable, safe and durable, easy to use.