ContactUsLinkComponent
VWR®, Clamps
VWR®, Clamps
Catalog # 241-0032
Supplier:  VWR Collection
undefined
VWR®, Clamps
Catalog # 241-0032
Supplier:  VWR Collection
Specifications
  • Spanning width:
    22 - 70 mm
  • Pk:
    2
Specifications
About this item
Laboral aluminium alloy.
  • Round jaws covered in vinyl to protect clamped items
  • Two easy action knurled screws for adjusting each of the movable clamping jaws independently of each other