Order Entry
Austria
ContactUsLinkComponent
VWR®, Inoculating Needles, Disposable
VWR®, Inoculating Needles, Disposable
Catalog # 612-2655
Supplier:  VWR Collection
undefined
VWR®, Inoculating Needles, Disposable
Catalog # 612-2655
Supplier:  VWR Collection

Specifications

  • Ø:
    1,45 mm
  • Colour:
    Violet
  • Description:
    Inoculating needles
  • Length:
    200 mm
  • Pk:
    1.000

Specifications

About this item

PS, sterile. Disposable plastic inoculating needles.

Packaging: Resealable pouches.