Inoculating loops and needles, disposable
Frequently Bought Together
Inoculating loops and needles, disposable
: Greiner Bio-One
VWR®, Precision Thermometers
: VWR Collection
Dehydrated culture media
: Merck
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Membrane filters with grid
: Sartorius
Hydrogen peroxide 6% (w/v) (20 volumes), GPR RECTAPUR®
: VWR Chemicals
Base materials
: Merck
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
Inoculating loops and needles, Ino-Loop™
: Simport Scientific
pH Buffer solutions
: Hach
Sodium hypochlorite solution 5% (w/w), Technical Grade
: PanReac AppliChem
tert-Butanol, anhydrous ≥99.5%
: Thermo Fisher Scientific