ContactUsLinkComponent
Inoculating loops with handle
Inoculating loops with handle
Catalog # 631-7144
Supplier:  HAMMACHER KARL
undefined
Inoculating loops with handle
Catalog # 631-7144
Supplier:  HAMMACHER KARL
Specifications
  • Pk:
    1
Specifications
About this item
Stainless steel 18/10, anti-magnetic. Spatula-shaped end.
Loop Ø: 4 mm
Length: 210 mm